Product Details
- SNP ID
-
rs4222142
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:4780355 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTTATGAACTGCTGTTGAATTCA[T/C]AGAACAGAATTTTTTACTGCCGCTG
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Mrpl15
PubMed Links
Gene Details
- Gene
- Mrpl15
- Gene Name
- mitochondrial ribosomal protein L15
View Full Product Details