Product Details
- SNP ID
-
rs3089638
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:34321951 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCCATTCAGCAAACCTGCTTGCA[T/C]ACTGTTTTTCCCACTCTCCAAATCT
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pbx3
PubMed Links
Gene Details
- Gene
- Pbx3
- Gene Name
- pre B cell leukemia homeobox 3
There are no transcripts associated with this gene.
View Full Product Details