Product Details

SNP ID
rs4223564
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:153009634 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGAGTTGGGAAGTCCTACCACAGGT[C/T]CCCCATTCTTCTGTGAGTCTCATCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Pdrg1 PubMed Links

Gene Details

Gene
Pdrg1
Gene Name
p53 and DNA damage regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178939.2 485 UTR 3 NP_849270.1
Gene
Ttll9
Gene Name
tubulin tyrosine ligase-like family, member 9
There are no transcripts associated with this gene.

View Full Product Details