Product Details

SNP ID
rs3666764
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:8757911 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAGTATAGAGTTAAAGTGCTGCAGA[G/A]AACACGATGCCTTGGAGATAGCTGT
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
Chd7 PubMed Links

Gene Details

Gene
Chd7
Gene Name
chromodomain helicase DNA binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277149.1 Intron NP_001264078.1

View Full Product Details