Product Details

SNP ID
rs4228405
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:93265504 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACTCGTTGCGGCTGGCACTTTTGAG[A/G]GAGGACAAGCCGTGCCTGTGGACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Elk3 PubMed Links

Gene Details

Gene
Elk3
Gene Name
ELK3, member of ETS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282967.1 794 Intron NP_001269896.1
NM_013508.2 794 Silent Mutation NP_038536.2
Gene
Gm32604
Gene Name
predicted gene, 32604
There are no transcripts associated with this gene.

View Full Product Details