Product Details
- SNP ID
-
rs3720676
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:33033808 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTTAATTACGAGATTTACCTCT[A/G]AATTGTTCAGAGGAAAAGTATTGTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Sdc2
PubMed Links
Gene Details
- Gene
- Sdc2
- Gene Name
- syndecan 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008304.2 |
2400 |
UTR 3 |
|
|
NP_032330.1 |
View Full Product Details