Product Details

SNP ID
rs3657951
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.19:24416601 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGCAGCTGTTTGTGTGACGAC[G/A]GCAATGGCACAGAGTCCCTTTGTTC
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
Pip5k1b PubMed Links

Gene Details

Gene
Pip5k1b
Gene Name
phosphatidylinositol-4-phosphate 5-kinase, type 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008846.2 Intron NP_032872.1

View Full Product Details