Product Details

SNP ID
rs4223998
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:79116294 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGCACATACATATATGTAGGTAAAA[C/T]ACCCACACACATTTTAAAAATTTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Rapgef2 PubMed Links

Gene Details

Gene
Rapgef2
Gene Name
Rap guanine nucleotide exchange factor (GEF) 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099624.2 Intron NP_001093094.2

View Full Product Details