Product Details
- SNP ID
-
rs3023032
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:152161462 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAAGTGGGAGGATTTGTGAGCCA[C/G]GGTCACTCCAAGTCCATGGGGATTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Espn
PubMed Links
Gene Details
- Gene
- Espn
- Gene Name
- espin
There are no transcripts associated with this gene.
- Gene
- Hes2
- Gene Name
- hairy and enhancer of split 2 (Drosophila)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008236.4 |
1514 |
UTR 3 |
|
|
NP_032262.2 |
View Full Product Details