Product Details

SNP ID
rs4225559
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:144248601 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCACCAGATCCCCCTCCCACACCCT[A/C]CTTAGGAAACATTTATATACTGTAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
2900089D17Rik PubMed Links

Gene Details

Gene
2900089D17Rik
Gene Name
RIKEN cDNA 2900089D17 gene
There are no transcripts associated with this gene.

Gene
Bri3
Gene Name
brain protein I3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163709.1 Intron NP_001157181.1
NM_018772.4 Intron NP_061242.3

View Full Product Details