Product Details

SNP ID
rs3023162
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.7:129293014 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGTTCAAGGTATTCTTGTAGG[A/G]TGTCTCAGGGTGTTTTAAAGTCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ppapdc1a PubMed Links

Gene Details

Gene
Ppapdc1a
Gene Name
phosphatidic acid phosphatase type 2 domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080963.1 Intron NP_001074432.1

View Full Product Details