Product Details
- SNP ID
-
rs4229294
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:26307605 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGTATACAAAATAAGTCCTCCCT[G/T]TTTTTTCTAATCCCTTAATCTTTAA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Rnf144a
PubMed Links
Gene Details
- Gene
- Rnf144a
- Gene Name
- ring finger protein 144A
View Full Product Details