Product Details
- SNP ID
-
rs4229496
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:91537097 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGAATATCAAAATAAATAACTTG[C/T]ATATCTTCCCAAGGGTCAAGGGGAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tshr
PubMed Links
Gene Details
- Gene
- Tshr
- Gene Name
- thyroid stimulating hormone receptor
View Full Product Details