Product Details

SNP ID
rs3091087
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:33186709 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCACACTGGGAACTGACGTTC[G/T]ATCAGAGGTTTCTCCAGGCTGTTAG
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
Camk4 PubMed Links

Gene Details

Gene
Camk4
Gene Name
calcium/calmodulin-dependent protein kinase IV
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009793.3 3273 UTR 3 NP_033923.2

View Full Product Details