Product Details
- SNP ID
-
rs3023938
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:59222187 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGTTTGTTTTGGTTTTTGGAATG[A/G]TAATGCTATTCTGTGATGTTGTGTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Chsy3
PubMed Links
Gene Details
- Gene
- Chsy3
- Gene Name
- chondroitin sulfate synthase 3
View Full Product Details