Product Details
- SNP ID
-
rs4222624
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:131667940 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTGGGATTATAGTGAAAATCTGT[A/G]TGCCTATTTCCCCAGGAGGCTGAGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ctse
PubMed Links
Gene Details
- Gene
- Ctse
- Gene Name
- cathepsin E
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007799.3 |
|
Intron |
|
|
NP_031825.2 |
View Full Product Details