Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STON1
Entrez Gene ID
102138016
Gene Name
stonin 1
Gene Aliases
-
Chromosome Location
Chr.13: 61675036 - 61743416 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005575923.2 XP_005575980.1 4 - 5 2478 78
XM_005575924.2 XP_005575981.1 3 - 4 2204 78

View Full Product Details