Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSHZ1
Entrez Gene ID
102127458
Gene Name
teashirt zinc finger homeobox 1
Gene Aliases
-
Chromosome Location
Chr.18: 4831415 - 4912330 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005586425.1 XP_005586482.1 1 - 2 76 72

View Full Product Details