Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MNT
Entrez Gene ID
102126184
Gene Name
MAX network transcriptional repressor
Gene Aliases
-
Chromosome Location
Chr.16: 2293315 - 2310758 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005582481.2 XP_005582538.1 2 - 3 1029 90

View Full Product Details