Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE2
Entrez Gene ID
102115013
Gene Name
jade family PHD finger 2
Gene Aliases
PHF15
Chromosome Location
Chr.6: 133578940 - 133637170 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005557795.2 XP_005557852.1 1 - 2 361 126
XM_015451896.1 XP_015307382.1 1 - 2 361 126

View Full Product Details