Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTURN
Entrez Gene ID
102146892
Gene Name
maturin, neural progenitor differentiation regulator homolog
Gene Aliases
C3H7orf41
Chromosome Location
Chr.3: 90498572 - 90527744 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005549889.2 XP_005549946.1 2 - 3 494 88

View Full Product Details