Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXC10
Entrez Gene ID
102133019
Gene Name
homeobox C10
Gene Aliases
-
Chromosome Location
Chr.11: 52886687 - 52891846 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005571049.2 XP_005571106.1 1 - 2 904 65

View Full Product Details