Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLRC1
Entrez Gene ID
102121106
Gene Name
killer cell lectin like receptor C1
Gene Aliases
nkg2A
Chromosome Location
Chr.11: 10790440 - 10804294 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284177.1 NP_001271106.1 1 - 2 196 74
XM_015430169.1 XP_015285655.1 2 - 3 1355 74
XM_015430170.1 XP_015285656.1 3 - 4 1798 74
XM_015430171.1 XP_015285657.1 2 - 3 1355 74
XM_015430172.1 XP_015285658.1 3 - 4 1798 74
GenBank mRNA AJ585528.1 - 1 - 2 196 74

View Full Product Details