Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAGLN
Entrez Gene ID
102120534
Gene Name
transgelin
Gene Aliases
-
Chromosome Location
Chr.14: 113153382 - 113158760 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283712.1 NP_001270641.1 2 - 3 260 63
GenBank mRNA AB220378.1 - 2 - 3 260 63

View Full Product Details