Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX60L
Entrez Gene ID
102140010
Gene Name
DExD/H-box 60 like
Gene Aliases
-
Chromosome Location
Chr.5: 167597872 - 167772301 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005556259.2 XP_005556316.1 3 - 4 458 100
XM_005556260.2 XP_005556317.1 2 - 3 311 100
XM_015451012.1 XP_015306498.1 3 - 4 431 100
XM_015451013.1 XP_015306499.1 4 - 5 633 100
XM_015451014.1 XP_015306500.1 2 - 3 338 100
XM_015451017.1 XP_015306503.1 3 - 4 481 100
XM_015451018.1 XP_015306504.1 3 - 4 483 100

View Full Product Details