Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIEF1
Entrez Gene ID
102118300
Gene Name
mitochondrial elongation factor 1
Gene Aliases
SMCR7L
Chromosome Location
Chr.10: 11656255 - 11672684 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005567222.2 XP_005567279.1 2 - 3 569 88

View Full Product Details