Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL10
Entrez Gene ID
102136121
Gene Name
BCL10 immune signaling adaptor
Gene Aliases
-
Chromosome Location
Chr.1: 141971727 - 141982887 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005542846.2 XP_005542903.1 2 - 3 886 148

View Full Product Details