Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED3
Entrez Gene ID
102117891
Gene Name
transmembrane p24 trafficking protein 3
Gene Aliases
-
Chromosome Location
Chr.7: 57773580 - 57784317 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005560228.2 XP_005560285.1 2 - 3 602 88

View Full Product Details