Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EFNA3
Entrez Gene ID
102133712
Gene Name
ephrin A3
Gene Aliases
-
Chromosome Location
Chr.1: 96715612 - 96724656 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541611.2 XP_005541668.1 2 - 3 556 59
XM_005541612.2 XP_005541669.1 2 - 3 556 59

View Full Product Details