Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFV3
Entrez Gene ID
102117222
Gene Name
NADH:ubiquinone oxidoreductase subunit V3
Gene Aliases
-
Chromosome Location
Chr.3: 3868494 - 3884316 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005548598.2 XP_005548655.1 2 - 3 233 135

View Full Product Details