Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FTMT
Entrez Gene ID
102130236
Gene Name
ferritin mitochondrial
Gene Aliases
-
Chromosome Location
Chr.6: 120807635 - 120808588 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005557601.2 XP_005557658.1 1 - 1 755 102

View Full Product Details