Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SP8
Entrez Gene ID
102145393
Gene Name
Sp8 transcription factor
Gene Aliases
-
Chromosome Location
Chr.3: 99960378 - 99966006 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015447645.1 XP_015303131.1 1 - 2 206 64

View Full Product Details