Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC6A
Entrez Gene ID
102120713
Gene Name
C-type lectin domain containing 6A
Gene Aliases
-
Chromosome Location
Chr.11: 8678529 - 8702738 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570039.2 XP_005570096.1 4 - 5 554 74

View Full Product Details