Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT7A
Entrez Gene ID
102137321
Gene Name
Wnt family member 7A
Gene Aliases
-
Chromosome Location
Chr.2: 144880829 - 144938045 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005547886.2 XP_005547943.1 2 - 3 583 122

View Full Product Details