Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXL7
Entrez Gene ID
102120523
Gene Name
F-box and leucine rich repeat protein 7
Gene Aliases
-
Chromosome Location
Chr.6: 15937588 - 16374018 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005556610.2 XP_005556667.1 2 - 3 451 103

View Full Product Details