Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STEAP1
Entrez Gene ID
102133382
Gene Name
STEAP family member 1
Gene Aliases
-
Chromosome Location
Chr.3: 121823740 - 121834145 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005550287.1 XP_005550344.1 4 - 5 961 113
XM_015447808.1 XP_015303294.1 5 - 6 875 113

View Full Product Details