Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ID1
Entrez Gene ID
102139470
Gene Name
inhibitor of DNA binding 1, HLH protein
Gene Aliases
-
Chromosome Location
Chr.10: 62923644 - 62924849 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005568627.2 XP_005568684.1 1 - 2 537 81

View Full Product Details