Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL17C
Entrez Gene ID
102143386
Gene Name
interleukin 17C
Gene Aliases
-
Chromosome Location
Chr.20: 77073934 - 77076591 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005592768.2 XP_005592825.1 2 - 3 377 139

View Full Product Details