Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
P2RY4
Entrez Gene ID
102131246
Gene Name
pyrimidinergic receptor P2Y4
Gene Aliases
-
Chromosome Location
Chr.X: 67288226 - 67291789 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015444352.1 XP_015299838.1 1 - 1 484 110

View Full Product Details