Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCUN1D3
Entrez Gene ID
102129304
Gene Name
defective in cullin neddylation 1 domain containing 3
Gene Aliases
-
Chromosome Location
Chr.20: 20144110 - 20190361 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591421.2 XP_005591478.1 1 - 2 131 83

View Full Product Details