Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFP62
Entrez Gene ID
102125639
Gene Name
ZFP62 zinc finger protein
Gene Aliases
-
Chromosome Location
Chr.6: 180859938 - 180877567 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558822.2 XP_005558879.1 1 - 2 210 66

View Full Product Details