Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SBK2
Entrez Gene ID
102134036
Gene Name
SH3 domain binding kinase family member 2
Gene Aliases
-
Chromosome Location
Chr.19: 56369751 - 56377119 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590407.2 XP_005590464.1 1 - 2 107 53

View Full Product Details