Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBTD2
Entrez Gene ID
102146576
Gene Name
ubiquitin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 171823317 - 171904981 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558526.2 XP_005558583.1 1 - 2 475 95

View Full Product Details