Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMM2
Entrez Gene ID
102146519
Gene Name
phosphomannomutase 2
Gene Aliases
-
Chromosome Location
Chr.20: 9093323 - 9140092 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285098.1 NP_001272027.1 1 - 2 111 86
XM_015442172.1 XP_015297658.1 1 - 2 86
XM_015442173.1 XP_015297659.1 1 - 2 86
GenBank mRNA AB125191.1 - 1 - 2 111 86

View Full Product Details