Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLFNL1
Entrez Gene ID
101925070
Gene Name
schlafen like 1
Gene Aliases
-
Chromosome Location
Chr.1: 186679875 - 186689660 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285026.1 NP_001271955.1 3 - 4 1256 95
XM_005543781.2 XP_005543838.1 4 - 5 5342 95
XM_005543782.2 XP_005543839.1 3 - 4 5362 95
XM_005543783.2 XP_005543840.1 3 - 4 5270 95
XM_005543784.2 XP_005543841.1 4 - 5 5277 95
XM_005543785.2 XP_005543842.1 4 - 5 5343 95
XM_005543786.2 XP_005543843.1 4 - 5 5341 95
XM_005543789.2 XP_005543846.1 5 - 6 5205 95
XM_005543790.2 XP_005543847.1 5 - 6 5232 95
XM_015451487.1 XP_015306973.1 3 - 4 1739 95
XM_015451491.1 XP_015306977.1 2 - 3 1824 95
XM_015451495.1 XP_015306981.1 5 - 6 5167 95
GenBank mRNA AB179349.1 - 3 - 4 1256 95

View Full Product Details