Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRPK2
Entrez Gene ID
101867513
Gene Name
SRSF protein kinase 2
Gene Aliases
-
Chromosome Location
Chr.3: 137533959 - 137836414 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285355.1 NP_001272284.1 10 - 11 1251 119
XM_015447283.1 XP_015302769.1 9 - 10 1195 119
XM_015447284.1 XP_015302770.1 9 - 10 2437 119
XM_015447285.1 XP_015302771.1 9 - 10 1558 119
XM_015447286.1 XP_015302772.1 10 - 11 1259 119
XM_015447287.1 XP_015302773.1 9 - 10 1196 119
XM_015447288.1 XP_015302774.1 9 - 10 1194 119
XM_015447289.1 XP_015302775.1 9 - 10 1195 119
XM_015447290.1 XP_015302776.1 9 - 10 1257 119
XM_015447292.1 XP_015302778.1 9 - 10 1195 119
GenBank mRNA AB168194.1 - 10 - 11 1251 119

View Full Product Details