Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRPK2
Entrez Gene ID
101867513
Gene Name
SRSF protein kinase 2
Gene Aliases
-
Chromosome Location
Chr.3: 137533959 - 137836414 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285355.1 NP_001272284.1 15 - 16 2107 83
XM_015447288.1 XP_015302774.1 14 - 15 2050 83
XM_015447290.1 XP_015302776.1 14 - 15 2113 83
GenBank mRNA AB168194.1 - 15 - 16 2107 83

View Full Product Details