Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA21
Entrez Gene ID
101925365
Gene Name
spermatogenesis associated 21
Gene Aliases
QtsA-16425
Chromosome Location
Chr.1: 211970895 - 212026561 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285317.1 NP_001272246.1 11 - 12 2010 117
XM_005544665.2 XP_005544722.1 11 - 12 2079 117
GenBank mRNA AB071053.1 - - 1367 117
AB168993.1 - 11 - 12 2010 117

View Full Product Details