Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDX5
Entrez Gene ID
101926602
Gene Name
DEAD-box helicase 5
Gene Aliases
-
Chromosome Location
Chr.16: 61838751 - 61845989 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284989.1 NP_001271918.1 5 - 6 697 74
XM_005584713.2 XP_005584770.1 4 - 5 624 74
XM_015437842.1 XP_015293328.1 5 - 6 777 74
XM_015437843.1 XP_015293329.1 3 - 4 457 74
GenBank mRNA AB169157.1 - 5 - 6 697 74
AB169223.1 - 3 - 4 449 74
AB170341.1 - 4 - 5 615 74

View Full Product Details