Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL30
Entrez Gene ID
101926789
Gene Name
mitochondrial ribosomal protein L30
Gene Aliases
L30mt, MRP-L30
Chromosome Location
Chr.13: 3462060 - 3489399 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284082.1 NP_001271011.1 3 - 4 210 106
XM_005575061.2 XP_005575118.1 4 - 5 390 106
XM_015433185.1 XP_015288671.1 4 - 5 416 106
XM_015433186.1 XP_015288672.1 3 - 4 176 106
GenBank mRNA AB169083.1 - 3 - 4 210 106

View Full Product Details